Trajan Graphite Short Reducing Ferrule With 0.8 Mm Hole For Detector/Injectors (Not For Msd Interface Connections), Pk10
Representative image only.
Trajan Graphite Short Reducing Ferrule With 0.8 Mm Hole For Detector/Injectors (Not For Msd Interface Connections), Pk10
Trajan | Mfr: 072636| Item: 1600747
Recommended applications are column to injector and column to FID connections. Unsuitable for connecting columns to mass spectrometers
$65.27CAD / Pkg of 10
$65.27CAD / Pkg of 10
Product Description
Key Features
  • Easy to use
  • Forms a reliable seal
  • Reusable; soft material; forms a soft grip to the capillary column
  • 6 month warranty
More About this Item
Variety of hole sizes available for 1/16", 1/8" and 1/4" fittings. 450°C temperature limit.
6 Month Warranty
Specifications
  • DescriptionTrajan Graphite short reducing ferrule with 0.8 mm hole for detector/injectors (not for MSD interface connections), PK10