Traceable® Big-Digit See-Thru Digital Thermometers with Calibration
Monitors temperature continuously
  • Automatically clears and updates MIN/MAX readings daily
  • Easy-to-read large, 1-1/8"H digits
Attractive transparent body easily attaches to a window or piece of glass. Use the thermometer to monitor temperature in storerooms, control rooms, fume hoods, laboratories, cleanrooms, refrigerators, plant areas, or outdoors. Meter simultaneously displays current temperature and daily min/max readings on a large, 1-1/8" high digit LCD; min/max readings are automatically cleared and updated daily.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
$62.03 - $62.91CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Meter Measurement Type
Availability Pricing
Traceable Big-Digit See-Thru Digital Thermometer; Fahrenheit 9008010 Mfr # 4159TR
COMPARE
Digital Indicator; Fahrenheit
$62.91CAD / Each
Traceable Big-Digit See-Thru Digital Thermometer; Celsius 9008011 Mfr # 4160TR
COMPARE
Digital Indicator; Celsius
$62.03CAD / Each