Torbal FC ZEUS Precision Pro Series Digital Force Gauges
Built in Wireless data transmission
  • Read results from any angle with large auto-rotate LCD display
  • Removable MicroSD card increases available memory
  • Wireless transmitter for cable free connection to PC, Laptop, or tablet
  • Onscreen graphing and histogram for visual reference of results
The FC ZEUS force gauge is ideal for advanced tension and compression applications which require speed and precision. Packed with functions and features including Peak mode, Fill Meter, Pass/Fail, Multi-Peak Mode, Peak Sensitivity, and Standard mode help increase productivity during force measurement testing. Not only strong and durable, but compact, ergonomic, and perfectly balanced to ensure comfort and stability while in your hand. Easily connect peripherals and store review data with numerous connectivity options including USB, RS-232, wireless, and I/O interface.
$1,536.75 - $2,818.60CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Sampling Rate
Availability Pricing
Torbal FC20 ZEUS Precision Pro Series Digital Force Gauge with Bluetooth, 4 lb/2 kg/20 N Capacity 2530451 Mfr # FC20
COMPARE
2 4 1000Hz
$1,536.75CAD / Each
Torbal FC200 ZEUS Precision Pro Series Digital Force Gauge with Bluetooth, 40 lb/20 kg/200 N Capacity 2530453 Mfr # FC200
COMPARE
20 40 1000Hz
$1,572.11CAD / Each
Torbal FC500 ZEUS Precision Pro Series Digital Force Gauge with Bluetooth, 100 lb/50 kg/500 N Capacity 2530454 Mfr # FC500
COMPARE
50 100 1000Hz
$1,623.68CAD / Each
Torbal FC20k ZEUS Precision Pro Series Digital Force Gauge with Bluetooth, 4000 lb/2000 kg/20 kN Capacity 2530459 Mfr # FC20k
COMPARE
2000 4000 1000Hz
$2,818.60CAD / Each