Torbal FB THOR Precision Series Digital Force Gauges
Affordable everyday force measurement
  • Large backlit LCD automatically adjusts to the position of the gauge
  • Keypad is simple to use and sealed to keep dirt and grime out
  • Multiple interfaces for connectivity
The FB THOR gauge offers affordability and scalability for everyday force measurements. Loaded with essential features and multiple modes and applications, these gauges are ideal for any tension and compression application. These gauges are designed for the most demanding applications. They are durable, compact, ergonomic, and perfectly balanced to ensure stability and comfort while in your hand. Easy PC connectivity with the USB or RS-232 interface and removable microSD memory means all data can be stored and easily transferred to a PC.
$1,193.45 - $2,562.23CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Sampling Rate
Availability Pricing
Torbal FB20 THOR Precision Series Digital Force Gauge, 4 lb/2 kg/20 N Capacity 2530463 Mfr # FB20
COMPARE
2 4 80Hz
$1,193.45CAD / Each
Torbal FB50 THOR Precision Series Digital Force Gauge, 10 lb/5 kg/50 N Capacity 2530464 Mfr # FB50
COMPARE
5 10 80Hz
$1,210.83CAD / Each
Torbal FB200 THOR Precision Series Digital Force Gauge, 40 lb/20 kg/200 N Capacity 2530465 Mfr # FB200
COMPARE
20 40 80Hz
$1,228.22CAD / Each
Torbal FB500 THOR Precision Series Digital Force Gauge, 100 lb/50 kg/500 N Capacity 2530466 Mfr # FB500
COMPARE
50 100 80Hz
$1,244.87CAD / Each
Torbal FB5k THOR Precision Series Digital Force Gauge, 1000 lb/500 kg/5 kN Capacity 2530469 Mfr # FB5k
COMPARE
500 1000 80Hz
$1,880.05CAD / Each
Torbal FB10k THOR Precision Series Digital Force Gauge, 2000 lb/1000 kg/10 kN Capacity 2530470 Mfr # FB10k
COMPARE
1000 2000 80Hz
$2,049.49CAD / Each
Torbal FB50k THOR Precision Series Digital Force Gauge, 10,000 lb/5000 kg/50 kN Capacity 2530472 Mfr # FB50k
COMPARE
5000 10000 80Hz
$2,562.23CAD / Each
Showing 1- 7 of 7