Thermo Scientific Super-Nuova Multiposition Stirring Hot Plates
  • Precise adjustment of settings in 1°/1-rpm increments displayed for each position on bright LED display
  • Flashing, red “hot top'' light provides visual warning to prevent accidental burns
Thermo Scientific Super-Nuova® units feature a powerful motor for quick startup without lurching. Convenient timer shuts off the unit when the timed cycle is complete — allows for unattended use and prevents accidental overheating. RS-232 port allows for output of time count, temperature, and set point directly to your PC.
$5,819.89 - $6,129.30CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Thermo Scientific Supernuova SP135935Q Multiposition Stirring Hot Plate, 4-position, 120V 8657680 Mfr # SP135935Q
COMPARE
120
$5,819.89CAD / Each
Thermo Scientific Supernuova SP135930-33Q Multiposition Stirring Hot Plate, 4-position, 240V 8657685 Mfr # SP135930-33Q
COMPARE
240
$6,129.30CAD / Each