Thermo Scientific Super-Nuova Multiposition Stirrers
Four individually controlled stirring positions
  • Precise adjustment of settings in 1°/1-rpm increments displayed for each position on bright LED display
  • Flashing, red “hot top'' light provides visual warning to prevent accidental burns
Thermo Scientific Super-Nuova® units feature a powerful motor for quick startup without lurching. Convenient timer shuts off the unit when the timed cycle is complete — allows for unattended use and prevents accidental overheating. RS-232 port allows for output of time count, temperature, and set point directly to your PC.
$4,464.37 - $4,626.44CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
No. Of Stirring Positions Power (VAC)
Availability Pricing
Thermo Scientific Supernuova S136035Q Multiposition Stirrer, 4-position, 120V 8657690 Mfr # S136035Q
COMPARE
4 120
$4,464.37CAD / Each
Thermo Scientific Supernuova S136030-33Q Multiposition Stirrer, 4-position, 240V 8657695 Mfr # S136030-33Q
COMPARE
4 240
$4,626.44CAD / Each