Thermo Scientific® Sterile Swabs
Individually wrapped to prevent sample contamination
  • Ideal for sample collection, cleaning, stirring, and microbial analysis
  • Sterile samplers provide accurate results
Obtain accurate results when collecting samples by using these sterile samplers. Choose from an assortment of sterile spatulas and swabs.
Wood spatulas are made from close-grained, northern white birch for a smooth, splinter-free finish. Spatulas have no taste or odor—perfect for spreading, stirring, and mixing food samples.
Rayon swabs have tips made of the finest USP grade of rayon fiber. They are disposable and come with either a wood or plastic shaft.
DACRON(R) swabs are inert and come with either a plastic or wood shaft. Swabs 14001-57 come with reusable transport tubes. Simply place the swabs back in the tube after use, then seal for transport to the lab.
Calcium alginate swabs have tips made of organic, biodegradable fiber. Select wood or plastic shaft.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (in) Description
Availability Pricing
Thermo Scientific Swab, Dacron, Sterile, Plastic Shaft, 6"L x 1/10" dia 1400110 Mfr # 149601J
COMPARE
6 Swab, Dacron, sterile, plastic shaft, 6"L x 1/10" dia
Call For Price
Thermo Scientific Wood Spatula, Splinter-Free Birch, Sterile, 6"L x 11/16"W 1400151 Mfr # 22025203
COMPARE
6 Wood Spatula, splinter-free birch, sterile, 6"L x 11/16"W