Thermo Scientific SAHARA Advanced Stainless Steel Heated Bath Circulators

Greater temperature stability for sensitive work
  • Microprocessor PID controller for fast and precise temperature control
  • Large selection of bath sizes to meet your application requirements
  • Control head can be indexed 90° left or right for easy viewing
SAHARA baths with Advanced (AC) Controllers have all the same features as the SC models with a larger
LCD, plus include a powerful three-speed force/suction pump for external open and closed loop applications.

Additional feature includes seven languages (English, German, French, Spanish, Italian, Chinese, and Japanese).
$6,335.58 - $10,253.32CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Reservoir Capacity (L) Description
Availability Pricing
Thermo Scientific AC150-S13 7-11L Heated Bath, Amb +13 to 150C, 115V 1214300 Mfr # 1551138
COMPARE
150 11 7-11L Heated Bath, Amb +13 to 150C, 115V
$6,335.58CAD / Each
Thermo Scientific AC200-S21 10-19L Heated Bath, Amb +13 to 200C, 115V 1214350 Mfr # 1561218
COMPARE
200 19 10-19L Heated Bath, Amb +13 to 200C, 115V
$7,795.71CAD / Each
Thermo Scientific AC150-S45 30-41L Heated Bath, Amb +13 to 150C, 115V 1214710 Mfr # 1551458
COMPARE
150 41 30-41L Heated Bath, Amb +13 to 150C, 115V
$9,048.09CAD / Each
Thermo Scientific AC200-S49 29-51L Heated Bath, Amb +13 to 200C, 115V 1214750 Mfr # 1561498
COMPARE
200 53 29-51L Heated Bath, Amb +13 to 200C, 115V
$10,253.32CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat