Thermo Scientific Orion pH Buffer Solutions
Common calibration values are color-coded for ease of use
  • Traceable to NIST standards
Each 475-mL (pint) buffer bottle comes color-coded for easy identification and packaged with buffer/temperature tables for accurate calibrations. Buffers are standardized against NIST-traceable pH references for superior accuracy.
$79.42 - $335.42CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Buffer Value Color
Availability Pricing
Thermo Scientific 910168 pH Buffer Solution, 1.68, Clear; 475 mL Bottle 5535028 Mfr # 910168
COMPARE
1.68 Clear
$82.22CAD / Each
Thermo Scientific 910104 pH Buffer Solution, 4.01, Pink; 475 mL Bottle 5535042 Mfr # 910104
COMPARE
4.01 Pink
$79.42CAD / Each
Thermo Scientific 910107 pH Buffer Solution, 7.00, Yellow; 475 mL Bottle 5535046 Mfr # 910107
COMPARE
7 Yellow
$83.91CAD / Each
Thermo Scientific 910110 pH Buffer Solution, 10.01, Blue; 475 mL Bottle 5535052 Mfr # 910110
COMPARE
10.01 Blue
$88.85CAD / Each
Thermo Scientific 810199 All-in-One pH Buffer Kit with Cleaning Solution 5535066 Mfr # 810199
COMPARE
Colored
$335.42CAD / Each
Thermo Scientific 910199 Standard All-in-One pH Buffer Kit 5535068 Mfr # 910199
COMPARE
Colored
$326.95CAD / Each