Thermo Scientific 910110 pH Buffer Solution, 10.01, Blue; 475 mL Bottle

Representative image only.

Thermo Scientific 910110 pH Buffer Solution, 10.01, Blue; 475 mL Bottle

Thermo Scientific | Mfr: 910110| Item: 5535052

Common calibration values are color-coded for ease of use

$88.85CAD / Each

$88.85CAD / Each

Buy It Together

Current Item

Thermo Scientific 910110 pH Buffer Solution, 10.01, Blue; 475 mL Bottle
Thermo Scientific 910110 pH Buffer Solution, 10.01, Blue; 475 mL Bottle
5535052

$88.85CAD / Each
+ =
Total Price 118.47 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Traceable to NIST standards

More About this Item

Each 475-mL (pint) buffer bottle comes color-coded for easy identification and packaged with buffer/temperature tables for accurate calibrations. Buffers are standardized against NIST-traceable pH references for superior accuracy.

Specifications
  • Buffer Value10.01
  • Buffer Kit Values10.01
  • Volume (mL)475
  • ColorBlue
  • DescriptionpH Buffer Solution, 10.01, Blue; 475 mL Bottle
Documents & Certificates
Reviews
Q&A