Thermo Scientific Nalgene Cryogenic Vial Holders
Polycarbonate vial holders features special design that locks the bottom of the vial in place
Polycarbonate vial holders allow single-hand opening/closing of cryogenic vials. Alphanumeric locating system is molded into the upper surface for easy sample identification. Rubber feet prevent sliding. Autoclavable.
$101.37 - $103.58CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubes Held Tube Size
Availability Pricing
Thermo Scientific Nalgene 5030-0510 Cryogenic vial holder for 50 (1.0 to 5.0 mL) vials RK-06754-90 Mfr # 5030-0510
COMPARE
50 1.0 to 5.0 mL
$101.37CAD / Each
Thermo Scientific Nalgene 5030-0505 Cryogenic vial holder for 25 (1.0 to 5.0 mL) vials RK-06754-92 Mfr # 5030-0505
COMPARE
25 1.0 to 5.0 mL
$103.58CAD / Each