Thermo Scientific Nalgene Benchtop Coolers

An excellent alternative to messy ice buckets for keeping reagents and enzymes cool
  • Lids feature a gridded write-on surface for sample identification
  • Stackable design saves valuable lab space
A convenient alternative to messy ice buckets. Body is molded from durable polycarbonate and filled with a nontoxic insulating gel; lid is made of clear polycarbonate (models 44001-10 and -25 have gel-filled white polycarbonate lid). Lids feature a gridded write-on surface for sample identification.

Models 44001-00, -10, and -25 include adapters to accommodate 0.5-mL tubes. Models 44001-20 and
-30 hold tubes up to 125-mm tall when lid is latched. All models have a stainless steel handle for a secure hold during transportation. Stackable design saves space.
$393.40 - $729.33CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubes Held Tube Size
Availability Pricing
Thermo Scientific Nalgene 5115-0012 Labtop cooler for 12 (0.5 to 2.0 mL) tubes RK-44001-00 Mfr # 5115-0012
COMPARE
12 0.5 to 2.0 mL
$393.40CAD / Each
Thermo Scientific Nalgene 5115-0032 Benchtop Cooler, -15C, 32 x 0.5-2.0 mL Tubes RK-44001-10 Mfr # 5115-0032
COMPARE
32 0.5 to 2.0 mL
$729.33CAD / Each
Thermo Scientific Nalgene 5116-1300 Labtop cooler for 12 (12 to 13 mm dia) tubes RK-44001-20 Mfr # 5116-1300
COMPARE
12 12 to 13 mm dia
$421.39CAD / Each
Thermo Scientific Nalgene 5116-1600 Labtop cooler for 12 (16 to 17 mm dia) tubes RK-44001-30 Mfr # 5116-1600
COMPARE
12 16 to 17 mm dia
$421.39CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat