Thermo Scientific Nalgene 5116-1600 Labtop cooler for 12 (16 to 17 mm dia) tubes

Representative image only.

Thermo Scientific Nalgene 5116-1600 Labtop cooler for 12 (16 to 17 mm dia) tubes

Thermo Scientific | Mfr: 5116-1600| Item: 4400130

An excellent alternative to messy ice buckets for keeping reagents and enzymes cool

$523.05CAD / Each

$523.05CAD / Each
Product Description

Key Features

  • Lids feature a gridded write-on surface for sample identification
  • Stackable design saves valuable lab space

More About this Item

A convenient alternative to messy ice buckets. Body is molded from durable polycarbonate and filled with a nontoxic insulating gel; lid is made of clear polycarbonate (models 44001-10 and -25 have gel-filled white polycarbonate lid). Lids feature a gridded write-on surface for sample identification.

Models 44001-00, -10, and -25 include adapters to accommodate 0.5-mL tubes. Models 44001-20 and
-30 hold tubes up to 125-mm tall when lid is latched. All models have a stainless steel handle for a secure hold during transportation. Stackable design saves space.

Specifications
  • Tubes Held12
  • Tube Size16 to 17 mm dia
  • Lowest Set Temperature (° C)1
  • Height (in)7 1/2
  • Height (cm)19.1
  • DescriptionLabtop cooler for 12 (16 to 17 mm dia) tubes
Reviews
Q&A