Thermo Scientific™ Hypersil™ BDS C18 Columns
Long life and trusted reproducibility for improved analyses
  • pH stable from 2 to 9
  • Base deactivating and endcapping reduce peak tailing for more precise results
Thermo Scientific™ Hypersil™ BDS (Base Deactivated Silica) columns have been trusted worldwide for over 20 years. Each column is manufactured with the same exactness and come with a certificate of analysis, ensuring you that each column will perform as promised. These high standards of manufacturing provide you with consistency and reliability year after year, meaning that you will not have to worry about changing procedures when you need to replace your column.
Hypersil BDS C18 Columns are economical, general-purpose columns that are perfect for QA and QC labs where reproducibility and long column life are required.
$1,364.06 - $2,600.53CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Thermo Scientific 28103-104630 BDS C18 Column - Length 100mm, 4.6mm ID, 3µm particle size 0188543 Mfr # 28103-104630
COMPARE
4.6 100 3
$1,598.63CAD / Each
Thermo Scientific Hypersil 28103-152130 BDS HPLC Column, C18, 3 µm, 150 mm L x 2.1 mm ID 0188544 Mfr # 28103-152130
COMPARE
2.1 150 3
$2,291.12CAD / Each
Thermo Scientific 28105-052130 BDS C18 Column - Length 50mm, 2.1mm ID, 5µm particle size 0188556 Mfr # 28105-052130
COMPARE
2.1 50 5
$1,364.06CAD / Each
Thermo Scientific 28105-154630 BDS C18 Column - Length 150mm, 4.6mm ID, 5µm particle size 0188571 Mfr # 28105-154630
COMPARE
4.6 150 5
$1,539.69CAD / Each
Thermo Scientific 28105-254030 BDS HPLC Column, C18, 5 µm, 250 mm L x 4.0 mm ID 0188578 Mfr # 28105-254030
COMPARE
4 250 5
$2,600.53CAD / Each
Thermo Scientific 28105-254630 BDS C18 Column - Length 250mm, 4.6mm ID, 5µm particle size 0188579 Mfr # 28105-254630
COMPARE
4.6 250 5
$1,620.73CAD / Each