Thermo Scientific Heratherm General Protocol Incubators
Ideal for standard applications in pharmaceutical, research, and food
  • Internal glass door allows sample viewing without disturbing temperature
  • Intuitive user interface for easy temperature setting
These gravity convection incubators provide top-quality temperature uniformity, ranging from 5°C above ambient to 75°C. A unique airflow design, advanced microprocessor control, and automatic overtemperature alarm ensure that an optimal environment is maintained.
Units are ideal for everyday heating and drying applications, operating up to 75°C with outstanding temperature stability. Features include a large, easy-to-read vacuum fluorescent display and corrosion-resistant stainless steel chamber with easy-to-clean rounded corners.
$4,788.52 - $18,071.13CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Thermo Scientific Heratherm IGS60 General Incubator, 2 cu ft, Gravity; 120V RK-38800-02 Mfr # 51028063
COMPARE
2.6 120
$4,788.52CAD / Each
Thermo Scientific Heratherm IGS400 General Incubator, 14.3 cu ft, Gravity; 120V RK-38800-03 Mfr # 51029321
COMPARE
14.3 120
$12,052.33CAD / Each
Thermo Scientific Heratherm IGS100 General Incubator, 3.5 cu ft, Gravity; 120V RK-38800-04 Mfr # 51028064
COMPARE
4 120
$5,245.27CAD / Each
Thermo Scientific Heratherm IGS750 General Incubator, 26.4 cu ft, Gravity; 120V RK-38800-05 Mfr # 51029333
COMPARE
26.4 120
$18,071.13CAD / Each
Thermo Scientific Heratherm IGS180 General Incubator, 6.3 cu ft, Gravity; 120V RK-38800-06 Mfr # 51028065
COMPARE
6.9 120
$7,028.07CAD / Each