Thermo Scientific Heratherm General Protocol (GP) Mechanical Convection Ovens
Optimally suited to everyday heating and drying applications up to 250°C
  • Energy efficient — up to 20% more energy savings compared to similar units
  • Small footprint saves space without compromising volume
  • Mechanical convection provides fast sample drying and heating with superior temperature uniformity
Energy-efficient mechanical convection ovens feature gentle air circulation to ensure accurate, even heating from 50 to 250°C, perfect for drying samples such as glassware and powders. Large easy-to-view vacuum fluorescent display with built-in timer and touch-button controls allows quick and simple temperature setting. The exhaust vent doubles as an access port for external temperature sensors.
Smooth corrosion-resistant galvanized steel inner chamber has rounded corners for easy cleaning. The door opens 180°, providing easy access to interior for cleaning and loading of probes. An automatic overtemperature alarm ensures your samples are always protected. The RS-232 interface lets you connect to your computer for documentation of parameters.
$5,186.33 - $7,175.41CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Thermo Scientific Heratherm 51028121 Heratherm GP Mechanical Oven; 2.3 cu ft/120V 3880036 Mfr # 51028121
COMPARE
2.3 120
$5,186.33CAD / Each
Thermo Scientific Heratherm 51028873 Heratherm GP Mechanical Oven; 3.65 cu ft/120V 3880037 Mfr # 51028873
COMPARE
3.75 120
$6,158.77CAD / Each
Thermo Scientific Heratherm 51028122 Heratherm GP Mechanical Oven; 3.65 cu ft/208-240V 3880038 Mfr # 51028122
COMPARE
3.75 208 / 240
$6,158.77CAD / Each
Thermo Scientific Heratherm 51028875 Heratherm GP Mechanical Oven; 6.3 cu ft/120V 3880039 Mfr # 51028875
COMPARE
6.3 120
$7,175.41CAD / Each
Thermo Scientific Heratherm 51028123 Heratherm GP Mechanical Oven; 6.3 cu ft/208-240V 3880040 Mfr # 51028123
COMPARE
6.3 208 / 240
$7,175.41CAD / Each