Thermo Scientific Heratherm Advanced Protocol (AP) Gravity Convection Ovens
Ideal for users requiring ultimate accuracy and heating capabilities up to 330°C
  • Gravity convection provides gentle drying with low air turbulence
  • Energy efficient-up to 20% more energy savings compared to similar units
  • Small footprint saves space without compromising volume
Advanced protocol gravity convection ovens feature a gentle air circulation pattern with low turbulence to ensure accurate, even heating from 50 to 330°C. Programmable controller with large easy-to-view display and touch-button controls can store up to 10 programs with 10 discrete steps. Unique timer allows you to automatically turn the oven on or off at a preset time or operate on a recurring weekly or hourly basis. Ovens feature an electronically controlled damper to vent the work space, a boost feature for rapid heating of cold oven, and a dedicated access port for the introduction of external temperature sensors.
Smooth stainless steel inner chamber has rounded corners for easy cleaning. The door opens 180°, providing easy access to interior for cleaning and loading of probes. An automatic overtemperature alarm ensures your samples are always protected. The RS-232 interface lets you connect to your computer for documentation of parameters.
$5,834.62 - $7,617.43CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Description
Availability Pricing
Thermo Scientific Heratherm 51028115 AP Gravity Convection Oven; 2.0 cu ft, 120V 3880042 Mfr # 51028115
COMPARE
2.15 AP Gravity Convection Oven; 2.0 cu ft, 120V
$5,834.62CAD / Each
Thermo Scientific Heratherm 51028117 AP Gravity Convection Oven; 6.1 cu ft, 208-240V 3880046 Mfr # 51028117
COMPARE
6.05 AP Gravity Convection Oven; 6.1 cu ft, 208-240V
$7,617.43CAD / Each