Thermo Scientific™ Finnpipette™ Novus Single Channel Electronic Pipettors
Ready to go out of the box!
  • So simple to use that you could skip reading the instruction manual
  • Graphical user interface
  • Lasting power and quick charging
  • Backlit digital display
  • 2-year warranty
  • Multi-language programming option
Intuitive menu and simple button layout, allow you to choose all the pipetting functions
you need quickly and easily. The large display is easy to read and shows the full names of
all functions. Easily accessible menu offers 10 different pipetting functions and 9 pipetting
speeds to handle a wide variety of pipetting applications. Select common functions such
as forward, reverse, diluting, and stepper pipetting or advanced functions like mixing,
sequential stepping, or program up to 9 of your own functions.
Solid-shaped and perfectly balanced handle fits comfortably and securely in your hand,
and together with the finger rest, it keeps pipetting stress and exertion to a minimum.
Ergonomically designed to let your index finger control dispensing with just a soft press
of the button. Adjust the finger rest a full 120° of movement to fit your hand exactly—
whether you’re left- or right-handed. Very low tip ejection force minimizes strain on your
thumb.
Graphical user interface in six languages: English, French, German, Italian, Spanish, and
Swedish. Easily switch languages to control and program the pipette in your preferred
language.
$2,093.69 - $2,139.36CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Code Max Volume (µL) Min Volume (µL)
Availability Pricing
Thermo Scientific Finnpipette 46200000 Electronic Pipette, 1-10 µL Micro-Tip, 100 to 240V 2499900 Mfr # 46200000
COMPARE
Pink 10 1
$2,093.69CAD / Each
Thermo Scientific Finnpipette 46200400 Electronic Pipette, 10-100 µL, 100 to 240V 2499904 Mfr # 46200400
COMPARE
Yellow 100 10
$2,139.36CAD / Each
Thermo Scientific Finnpipette 46200600 Electronic Pipette, 100-1000 µL, 100 to 240V 2499908 Mfr # 46200600
COMPARE
blue 1000 100
$2,098.11CAD / Each
Thermo Scientific Finnpipette 46200700 Electronic Pipette, 0.5-5mL, 100 to 240V 2499909 Mfr # 46200700
COMPARE
green 5000 500
$2,098.11CAD / Each
Thermo Scientific Finnpipette 46200800 Electronic Pipette, 1-10mL, 100 240V 2499910 Mfr # 46200800
COMPARE
red 10000 1000
$2,121.68CAD / Each