Thermo Scientific Cimarec+ Stirring Hot Plates
Advanced performance and exceptional safety in a user-friendly design
  • StirTrac™ control feature provides constant speed, strong magnetic coupling, and ultra-fast braking
  • Microprocessor temperature control produces precise and stable heat distribution
  • Prominent hot top indicator prevents accidental burns
  • Automatic overtemperature protection cuts power to avoid fires and other accidents
  • Optional accessories offer enhanced safety features
Cimarec+ stirring hot plates are ideal for academic and research institutions desiring reliable, safe, and easy operation. The hot top indicator remains lit as long as the plate temperature is above 50°C, even if the hot plate is turned off. Temperature is adjustable in one degree increments, allowing you to control the temperature more accurately. Optional external temperature probe (order separately) provides the feedback necessary to keep your sample at the desired temperature. Optional splash guard (order separately) protects you and your sample from mishaps. Optional rod and clamps (order separately) provide the means to secure your sample container to the top plate.
$917.92 - $1,421.82CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Top Plate Length (in) Top Plate Material
Availability Pricing
Thermo Scientific Cimarec Stirring Hot Plate, 4x4" Ceramic; 120 VAC RK-04600-05 Mfr # SP88854100
COMPARE
100 to 120 4 1/4 Ceramic
$917.92CAD / Each
Thermo Scientific Cimarec SP88854105 Stirring Hot Plate, 4x4" Ceramic; 230 VAC RK-04600-06 Mfr # SP88854105
COMPARE
230 4 1/4 Ceramic
$1,016.64CAD / Each
Thermo Scientific Cimarec Stirring Hot Plate, 7x7" Ceramic; 120 VAC RK-04600-12 Mfr # SP88857100
COMPARE
100 to 120 7 1/4 Ceramic
$1,012.22CAD / Each
Thermo Scientific Cimarec SP88857105 Stirring Hot Plate, 7x7" Ceramic; 230V EU RK-04600-13 Mfr # SP88857105
COMPARE
230 7 1/4 Ceramic
$1,091.78CAD / Each
Thermo Scientific Cimarec SP88857106 Stirring Hot Plate, 7x7" Ceramic; 230V AU/CN RK-04600-14 Mfr # SP88857106
COMPARE
230 7 1/4 Ceramic
$1,091.78CAD / Each
Thermo Scientific Cimarec Stirring Hot Plate, 7x7" Ceramic; 230V UK RK-04600-15 Mfr # SP88857107
COMPARE
230 7 1/4 Ceramic
$1,091.78CAD / Each
Thermo Scientific Cimarec Stirring Hot Plate, 7x7" Aluminum; 120 VAC RK-04600-21 Mfr # SP88857104
COMPARE
120 7 1/4 Aluminum
$1,301.00CAD / Each
Thermo Scientific Cimarec Stirring Hot Plate, 7x7" Aluminum; 230 VAC RK-04600-22 Mfr # SP88857108
COMPARE
230 7 1/4 Aluminum
$1,309.84CAD / Each
Thermo Scientific Cimarec Stirring Hot Plate, 10x10" Ceramic; 120 VAC RK-04600-44 Mfr # SP88850100
COMPARE
120 10 1/4 Ceramic
$1,421.82CAD / Each
Thermo Scientific Cimarec SP88850105 Stirring Hot Plate, 10x10" Ceramic; 230V EU RK-04600-45 Mfr # SP88850105
COMPARE
230 10 1/4 Ceramic
$1,421.82CAD / Each
Showing 1- 10 of 10