Thermo Scientific Cimarec Mini and Micro Stirrers
Ultra-compact works in the smallest spaces
  • No moving parts for maintenance- and wear-free operation
  • Ideal for system integration with analytical devices
  • Easy-to-use external remote controlller for safe operation
Mini and Mini 20 Models are as small as a penny, perfect for cuvette stirring. Their compact size makes them ideal for use with cuvette holders and photometers. Stir volumes up to 5 mL. The 04651-36 and -56 include a 7-watt controller and the 04656-60 includes a more powerful 20-watt controller for higher speeds.
Micro and Micro 20 Models are small enough to fit in your hand. Resistant to temperatures up to 95 °C in water and 120 °C in air for submersion in water baths, high humidity environments, and ovens. Their hermetically-sealed stainless steel housing is resistant to aggressive media, micro-organisms, and dust. Stir volumes up to 1 L. The 04651-44 and -54 include a 7-watt controller, while the 04651-55 includes a more powerful 20-watt controller for higher speeds.
All models feature an extremely flexible 0.3 mm thick foil cable to conserve on space.
$1,267.12 - $1,361.41CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) Power (VAC)
Availability Pricing
Thermo Scientific Cimarec 50088130 i Mini Stirrer with Telemodul Controller 230V Euro 0465154 Mfr # 50088130
COMPARE
1000 230
$1,361.41CAD / Each
Thermo Scientific Cimarec 50088150 Micro, with 7-watt controller, 115 V 0465156 Mfr # 50088150
COMPARE
1000 115
$1,267.12CAD / Each