Thermo Scientific Barnstead E-pure 3- and 4-Holder Water Purification Systems
Produce ASTM Type I water Recirculation pump ensures water purity
Thermo Scientific Barnstead E-pure™ systems meet or exceed ASTM, CAP, and NCCLS requirements for bacteria free, Type I reagent grade water. Inert wetted parts prevent contamination of purified water. A digital resistivity meter (included) indicates water purity. A built-in, whisper-quiet recirculation pump with pressure regulator ensures Type I reagent grade water on demand. Order low-water and low-pressure pump protectors separately.
Choose from wall mountable three- or four-holder purification systems. Order disposable cartridges (required) by searching our Replacement Cartridge section; upgrade your system by adding an optional Remote Dispenser.
$10,932.55 - $11,448.24CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Thermo Scientific Barnstead D4631 3-Holder Water Purification System; 120V 9925200 Mfr # D4631
COMPARE
120
$10,932.55CAD / Each
Thermo Scientific D4641 4-Holder Water Purification System; 120V 9925210 Mfr # D4641
COMPARE
120
$11,448.24CAD / Each