Thermo Scientific D4641 4-Holder Water Purification System; 120V

Representative image only.

Thermo Scientific D4641 4-Holder Water Purification System; 120V

Thermo Scientific | Mfr: D4641| Item: 9925210

Produce ASTM Type I water Recirculation pump ensures water purity

$11,448.24CAD / Each

$11,448.24CAD / Each

Buy It Together

Current Item

Thermo Scientific D4641 4-Holder Water Purification System; 120V
Thermo Scientific D4641 4-Holder Water Purification System; 120V
9925210

$11,448.24CAD / Each
+ =
Total Price 13923.54 ADD 2 ITEMS TO CART
Product Description

Key Features

  • 1 year warranty

More About this Item

Thermo Scientific Barnstead E-pure™ systems meet or exceed ASTM, CAP, and NCCLS requirements for bacteria free, Type I reagent grade water. Inert wetted parts prevent contamination of purified water. A digital resistivity meter (included) indicates water purity. A built-in, whisper-quiet recirculation pump with pressure regulator ensures Type I reagent grade water on demand. Order low-water and low-pressure pump protectors separately.
Choose from wall mountable three- or four-holder purification systems. Order disposable cartridges (required) by searching our Replacement Cartridge section; upgrade your system by adding an optional Remote Dispenser.

1 Year Warranty
Specifications
  • ASTM Water GradeType I
  • Max Flow Rate (L/min)2
  • Max Flow Rate (L/hour)120
  • Max Flow Rate (GPH)26.4
  • Number of Holders4
  • Inlet Port1/4" NPT(F)
  • SourceTap water
  • Feed Water Requirements (pressure)100
  • Feed Water Requirements (Temperature)0-49⁰C
  • Max Inlet Water Temperature (° F)120
  • Min Inlet Water Temperature (° F)40
  • Max Inlet Water Temperature (° C)49
  • Min Inlet Water Temperature (° C)4
  • Min Inlet Pressure (PSI)5
  • Max Inlet Pressure (PSI)100
  • Min Inlet Pressure (bar)0.3
  • Max Inlet Pressure (bar)6.9
  • Power (VAC)120
  • Height (in)28
  • Depth (in)7.5
  • Width (in)35
  • Height (cm)71.1
  • Depth (cm)19
  • Width (cm)88.9
  • Description4-Holder Water Purification System; 120V
  • Warranty1 year
Reviews
Q&A