Tempilaq Temperature-Indicating Liquid Paints
Easy confirmation that a target temperature has been met
  • Nonflammable for maximum safety
  • Easy application with quick-dry formula on any surface
Irreversible temperature-indicating liquids offer an easy, cost-effective method to verify a specific temperature on a wide range of surfaces under dynamic conditions. Paints are available in 43 different temperature ratings from 175 to 1900°F (79 to 1038°C) with accuracy within ±1% of rated temperature. They are ideal for heat treating, preventive maintenance, and any other temperature-sensitive applications. When applied to a dry smooth surface, the liquid quickly dries and forms a dull and opaque film. The film melts at the designated temperature, leaving a distinctly different mark to confirm the target temperature has been achieved. Available in 2-oz (59-mL) bottles.
$67.33 - $73.96CAD / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Temperature Points (° C) Temperature Points (° F)
Availability Pricing
Tempilaq Temperature-Indicating Paint, 300⁰F (2-oz Jar) RK-09036-62 Mfr # TEMPLAQ0300F2OZ
COMPARE
149 300
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 325⁰F (2-oz Jar) RK-09036-64 Mfr # TEMPLAQ0325F2OZ
COMPARE
163 325
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 350⁰F (2-oz Jar) RK-09036-65 Mfr # TEMPLAQ0350F2OZ
COMPARE
177 350
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 375⁰F (2-oz Jar) RK-09036-67 Mfr # TEMPLAQ0375F2OZ
COMPARE
191 375
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 400⁰F (2-oz Jar) RK-09036-69 Mfr # TEMPLAQ0400F2OZ
COMPARE
218 425
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 450⁰F (2-oz Jar) RK-09036-70 Mfr # TEMPLAQ0450F2OZ
COMPARE
232 450
$72.79CAD / Each
Tempilaq Temperature-Indicating Paint, 475⁰F (2-oz Jar) RK-09036-71 Mfr # TEMPLAQ0475F2OZ
COMPARE
246 475
$72.79CAD / Each
Tempilaq Temperature-Indicating Paint, 500⁰F (2-oz Jar) RK-09036-73 Mfr # TEMPLAQ0500F2OZ
COMPARE
260 500
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 650⁰F (2-oz Jar) RK-09036-78 Mfr # TEMPLAQ0650F2OZ
COMPARE
343 650
$71.31CAD / Each
Tempilaq Temperature-Indicating Paint, 700⁰F (2-oz Jar) RK-09036-79 Mfr # TEMPLAQ0700F2OZ
COMPARE
371 700
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 750⁰F (2-oz Jar) RK-09036-80 Mfr # TEMPLAQ0750F2OZ
COMPARE
399 750
$67.78CAD / Each
Tempilaq Temperature-Indicating Paint, 800⁰F (2-oz Jar) RK-09036-81 Mfr # TEMPLAQ0800F2OZ
COMPARE
427 800
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 850⁰F (2-oz Jar) RK-09036-82 Mfr # TEMPLAQ0850F2OZ
COMPARE
454 850
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 900⁰F (2-oz Jar) RK-09036-83 Mfr # TEMPLAQ0900F2OZ
COMPARE
482 900
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 950⁰F (2-oz Jar) RK-09036-84 Mfr # TEMPLAQ0950F2OZ
COMPARE
510 950
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 1000⁰F (2-oz Jar) RK-09036-85 Mfr # TEMPLAQ1000F2OZ
COMPARE
538 1000
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 1100⁰F (2-oz Jar) RK-09036-88 Mfr # TEMPLAQ1100F2OZ
COMPARE
593 1100
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 1150⁰F (2-oz Jar) RK-09036-89 Mfr # TEMPLAQ1150F2OZ
COMPARE
621 1150
$72.79CAD / Each
Tempilaq Temperature-Indicating Paint, 1200⁰F (2-oz Jar) RK-09036-90 Mfr # TEMPLAQ1200F2OZ
COMPARE
649 1200
$73.96CAD / Each
Tempilaq Temperature-Indicating Paint, 1300⁰F (2-oz Jar) RK-09036-92 Mfr # TEMPLAQ1300F2OZ
COMPARE
704 1300
$73.96CAD / Each
Showing 1 - 20 of 21