Tempilaq Temperature-Indicating Paint, 475⁰F (2-oz Jar)
Representative image only.
Tempilaq Temperature-Indicating Paint, 475⁰F (2-oz Jar)
Tempil | Mfr: TEMPLAQ0475F2OZ | Item: 0903671
Easy confirmation that a target temperature has been met
$89.73CAD / Each
$89.73CAD / Each
Buy It Together
Current Item
Tempilaq Temperature-Indicating Paint, 475⁰F (2-oz Jar)
Tempilaq Temperature-Indicating Paint, 475⁰F (2-oz Jar)
0903671
$89.73CAD / Each
+
Tempilstik Temperature Indicating Crayon, 100⁰F; 5/Pk
0903556
$169.44CAD / Pkg of 5
=
Total Price 259.17 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Nonflammable for maximum safety
  • Easy application with quick-dry formula on any surface
  • 1 year warranty
More About this Item
Irreversible temperature-indicating liquids offer an easy, cost-effective method to verify a specific temperature on a wide range of surfaces under dynamic conditions. Paints are available in 43 different temperature ratings from 175 to 1900°F (79 to 1038°C) with accuracy within ±1% of rated temperature. They are ideal for heat treating, preventive maintenance, and any other temperature-sensitive applications. When applied to a dry smooth surface, the liquid quickly dries and forms a dull and opaque film. The film melts at the designated temperature, leaving a distinctly different mark to confirm the target temperature has been achieved. Available in 2-oz (59-mL) bottles.
1 Year Warranty
Specifications
  • Number of Temperature Indicators1
  • Temperature Points (° F)475
  • Temperature Points (° C)246
  • DescriptionTemperature-Indicating Paint, 475⁰F (2-oz Jar)