Taylor Classic Series NSF-Stainless Steel Pocket Bimetal Thermometers
1” polycarbonate magnifying lens for easy reading
  • Factory calibrated to one point
  • Hermetically sealed case
  • Includes plastic storage sleeve with pocket clip
  • Sleeve includes built-in wrench for convenient field adjustment
  • Easy-to-read dial
This Taylor Precisio Classic Series instant read thermometer features a 1" high-visibility, graphic temperature dial with a shatter-proof plastic lens and a 5" stainless steel stem. Its exclusive protective pocket case has additives that inhibit the growth of bacteria and fungus. The pocket case also doubles as a calibration tool with a built in recalibration wrench.
$11.64 - $46.56CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Max Temperature (° F) Min Temperature (° F)
Availability Pricing
Taylor 6092N Instant Read 0 to 220°F Pocket Bimetal Thermometer 0808014 Mfr # 6092N (077784609217)
COMPARE
104 220 0
$14.14CAD / Each
Taylor 6093N Instant Read 50 to 550°F Pocket Bimetal Thermometer 0808015 Mfr # 6093N
COMPARE
288 550 50
$11.64CAD / Each
Taylor 6094N Instant Read -40 to 50°C Pocket Bimetal Thermometer 0808016 Mfr # 6094N
COMPARE
50 122 -40
$11.64CAD / Each
Taylor 6095N Instant Read 25 to 125°F Pocket Bimetal Thermometer 0808017 Mfr # 6075
COMPARE
52 125 25
$13.11CAD / Each
Taylor 6096N Instant Read -40 to 160°F Pocket Bimetal Thermometer 0808018 Mfr # 6096N
COMPARE
71 160 -40
$11.64CAD / Each
Taylor 6099N Instant Read -10 to 110°C Pocket Bimetal Thermometer 0808019 Mfr # 6099N
COMPARE
110 230 14
$11.64CAD / Each
Taylor 6084J-12 Instant Read 50 to 550°F Pocket Bimetal Thermometer with 12" Stem 9000043 Mfr # 6084J12
COMPARE
288 550 50
$46.56CAD / Each
Taylor 6091N  Instant Read -40 to 120°F Pocket Bimetal Thermometer 9000101 Mfr # 6091N
COMPARE
49 120 -40
$12.97CAD / Each
Showing 1- 8 of 8