Standard Paired Quartz Cuvettes for UV Spectroscopy
Quartz cuvettes can be used for the entire UV spectra down to 200 nm and have a guaranteed 80% transmission at 200 nm
  • 3.5 mL capacity
  • 10 mm pathlength
Cuvettes come in matched pairs in standard, semimicro, or micro size, and can be ordered with either a cover of PTFE resin or a stopper of PTFE resin. All cuvettes have a 10-mm pathlength. All pairs are accurately matched to give the same value for the same solution within 1% tolerance.
$492.11 - $671.87CAD / Pkg of 2
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Capacity (mL) Pathlength (mm)
Availability Pricing
COMPARE
3.5 10
$492.11CAD / Pkg of 2
COMPARE
3.5 10
$671.87CAD / Pkg of 2