Stainless Steel Electronic Calipers with Ceramic Capacitive Sensor
  • Unique ceramic capacitive sensor
  • Measure inside, outside, depth, step
  • Direct inch/metric conversion
  • 0.0005"/0.01 mm resolution
$503.90 - $741.85CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max range (in)
Availability Pricing
Fowler 54-100-000-1 Digital Caliper, SS with Ceramic Capacitive Sensor; 0-6 in. 9715208 Mfr # 54-100-000-2
COMPARE
6
$503.90CAD / Each
Fowler 54-100-112-1 12" Digital Caliper with LCD - +/-0.0015 in/+/-0.04 mm 9715212 Mfr # 54-100-112-2
COMPARE
12
$741.85CAD / Each