SHOWA NightHawk Black Nitrile Gloves
Feature natural antistatic properties
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
Powder-free, black nitrile gloves are designed to minimize the particulate and static generation that is often a concern in electronics or critical parts handling. Gloves are formulated without the accelerators commonly used in nitrile gloves — reduce allergic reaction in workers with sensitive skin. Ambidextrous.
$15.47 - $20.63CAD / Box of 50
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Size
Availability Pricing
SHOWA 7700PFT-S Black Nitrile Gloves, Small, Box of 50 RK-86485-00 Mfr # 7700PFT-S
COMPARE
9 1/2 Small
$15.62CAD / Box of 50
SHOWA 7700PFT-M Black Nitrile Gloves, Medium, Box of 50 RK-86485-01 Mfr # 7700PFT-M
COMPARE
9 1/2 Medium
$15.47CAD / Box of 50
SHOWA 7700PFT-L Black Nitrile Gloves, Large, Box of 50 RK-86485-02 Mfr # 7700PFT-L
COMPARE
9 1/2 Large
$15.47CAD / Box of 50
SHOWA 7700PFT-XL Black Nitrile Gloves, X-large, Box of 50 RK-86485-03 Mfr # 7700PFT-XL
COMPARE
9 1/2 X-Large
$20.63CAD / Box of 50