SHOWA 7700PFT-S Black Nitrile Gloves, Small, Box of 50
Representative image only.
SHOWA 7700PFT-S Black Nitrile Gloves, Small, Box of 50
SHOWA | Mfr: 7700PFT-S| Item: 8648500
Feature natural antistatic properties
$18.71CAD / Box of 50
BUY MORE & SAVE
  • 1-9$18.71 CAD / Box of 50
  • 10+$16.84 CAD / Box of 50
$18.71CAD / Box of 50
BUY MORE & SAVE
  • 1-9$18.71 CAD / Box of 50
  • 10+$16.84 CAD / Box of 50
Product Description
Key Features
  • Formulated without the accelerators
  • Ambidextrous
  • 1 year warranty
More About this Item
Powder-free, black nitrile gloves are designed to minimize the particulate and static generation that is often a concern in electronics or critical parts handling. Gloves are formulated without the accelerators commonly used in nitrile gloves — reduce allergic reaction in workers with sensitive skin.
1 Year Warranty
Specifications
  • Thickness (mil)4
  • MaterialNitrile
  • SizeSmall
  • Length (in)9 1/2
  • Length (cm)24
  • ColorBlack
  • ComplianceFDA 21 CFR 177.2600
  • SterileNo
  • DescriptionBlack Nitrile Gloves, Small, Box of 50