Shimpo Data-Logging Digtial Force Gauge
On board statistics provides immediate results and analysis of test data
  • High accuracy +/-0.2% F.S., ideal for strict tolerances
  • Measure and record compression and tension tests- permits true “push-pull” testing
  • Rugged, ergonomic die cast aluminum construction allows usage on production floor
  • Variable display response rate aids viewing readings
  • Quick retrieval of peak measurement with a single push button
  • Flexible user preference with one button selection of up to 4 engineering units
  • Dual power capable with rechargeable battery or AC line voltage
  • Field calibration through keypad eliminates down time
This series of force gauges is a versatile family of instruments perfect for quality inspections, finished goods testing, R&D, and other force related applications. These gauges cover ranges from a few ounces up to 200 lbs. A 180° reversible display and dual printed keypad allows usage as a portable gauge or mounted on a test stand. The 1000 Hz sample rate provides faster sampling, producing better resolution and more accurate results. One million operation load cell with superior overload protection provides a long-life, damage free product.
All models feature maximum data collection flexibility with three outputs: USB, RS-232, and analog. Onboard statistics calculations provide fast access to maximum, minimum, average, and standard deviation data. Additional features include 1,000-point continuous memory for reviewing data points and transferring to PC/Laptop, and a built-in Hi-Lo comparator function for easy ‘Go-No Go’ Testing.
$1,318.68 - $1,392.35CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (lbs)
Availability Pricing
Shimpo FGV-0.5XY Compact Gauge with USB, Analog, and RS-232 Output; 0.5 lb Capacity 9395131 Mfr # FGV-0.5XY
COMPARE
0.5
$1,318.68CAD / Each
Shimpo FGV-1XY Compact Gauge with USB, Analog, and RS-232 Output; 1 lb Capacity 9395132 Mfr # FGV-1XY
COMPARE
1
$1,318.68CAD / Each
Shimpo FGV-5XY Compact Gauge with USB, Analog, and RS-232 Output; 5 lb Capacity 9395134 Mfr # FGV-5XY
COMPARE
5
$1,318.68CAD / Each
Shimpo FGV-10XY Compact Gauge with USB, Analog, and RS-232 Output; 10 lb Capacity 9395135 Mfr # FGV-10XY
COMPARE
10
$1,318.68CAD / Each
Shimpo FGV-50XY Compact Gauge with USB, Analog, and RS-232 Output; 50 lb Capacity 9395137 Mfr # FGV-50XY
COMPARE
50
$1,318.68CAD / Each
Shimpo FGV-100XY Compact Gauge with USB, Analog, and RS-232 Output;  100 lb Capacity 9395138 Mfr # FGV-100XY
COMPARE
100
$1,318.68CAD / Each
Shimpo FGV-200XY Compact Gauge with USB, Analog, and RS-232 Output; 200 lb Capacity 9395139 Mfr # FGV-200XY
COMPARE
200
$1,392.35CAD / Each
Showing 1- 7 of 7