Semimicro Cuvettes with Clear Thick Wall
Matched pairs for less data variability
  • Paired cuvettes are matched to give the same value for the same solution within 1% tolerance
The glass cuvettes have % transmission guaranteed 82% at 320 nm. Tolerance will be within 1%. The quartz cuvette has 80% transmission guaranteed at 200 nm. Tolerance is within 1%
$373.14CAD / Pkg of 2
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Capacity (mL) Pathlength (mm)
Availability Pricing
COMPARE
Glass 1.4 10
$373.14CAD / Pkg of 2