Sartorius Sartobind® Lab IEX Ion Exchange Membrane Adsorber
High dynamic binding capacity—ideal for capturing large proteins and viruses
  • Ready-to-use reusable adsorbers optimize cost efficiency
  • Connect in series for increased binding capacity
Sartorius Sartobind® Lab membrane adsorbers for ion exchange chromatography are ready-to-use units in a syringe filter format. They are based on the patented macroporous Sartobind membranes, with anion or cation exchange ligands covalently bound to the entire internal surface of the pores. This results in a chromatography matrix with superior flow rates and high binding capacity. Covalent linkage of ion exchange ligands to the membrane is very stable, allowing for many cycles of re-use without loss of binding capacity. Equipped with standard Luer lock connectors, Sartobind Lab units enable simple, equipment-free handling by syringe, or can be easily connected to FPLC systems for automated purifications. The units are compatible with most common chromatography buffers and can be re-used > 1,000 times, ensuring high-cost efficiency.
Sartobind Lab units for ion exchange are available with strong (Q) or weak (D) anion and strong cation (S) exchangers, and in three bed volumes, 0.41, 2.1, and 2.8 mL, corresponding to dynamic binding capacities between 10.5 and 80 mg protein per unit.
$221.01 - $418.44CAD / Pkg of 2
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Effective Filtration Area (cm²) Filter Size
Availability Pricing
Sartorius Sartobind® Lab D Anion Exchange Membrane Adsorber, 2.1 mL; 2/PK 6482272 Mfr # 93IEXD42DB-12--V
COMPARE
2.1 75 25 mm Diameter Membrane
$318.25CAD / Pkg of 2
Sartorius Sartobind® Lab Q Anion Exchange Membrane Adsorber, 2.8 mL; Each 6482273 Mfr # 93IEXQ42BC-12
COMPARE
2.8 100 50 mm Diameter Membrane
$418.44CAD / Each
Sartorius Sartobind® Lab Q Anion Exchange Membrane Adsorber, 2.1 mL; 2/PK 6482274 Mfr # 93IEXQ42DB-12--V
COMPARE
2.1 75 25 mm Diameter Membrane
$318.25CAD / Pkg of 2
Sartorius Sartobind® Lab Q Anion Exchange Membrane Adsorber, 0.41 mL; 4/PK 6482275 Mfr # 93IEXQ42GB-12--A
COMPARE
0.41 15 25 mm Diameter Membrane
$226.90CAD / Pkg of 4
Sartorius Sartobind® Lab S Cation Exchange Membrane Adsorber, 2.8 mL; Each 6482276 Mfr # 93IEXS42BC-12
COMPARE
2.8 100 50 mm Diameter Membrane
$418.44CAD / Each
Sartorius Sartobind® Lab S Cation Exchange Membrane Adsorber, 2.1 mL; 2/PK 6482277 Mfr # 93IEXS42DB-12--V
COMPARE
2.1 75 25 mm Diameter Membrane
$318.25CAD / Pkg of 2
Sartorius Sartobind® Lab S Cation Exchange Membrane Adsorber, 0.41 mL; 4/PK 6482278 Mfr # 93IEXS42GB-12--A
COMPARE
0.41 15 25 mm Diameter Membrane
$221.01CAD / Pkg of 4
Showing 1- 7 of 7