Sartorius Sartobind® Lab Q Anion Exchange Membrane Adsorber, 0.41 mL; 4/PK

Representative image only.

Sartorius Sartobind® Lab Q Anion Exchange Membrane Adsorber, 0.41 mL; 4/PK

Sartorius | Mfr: 93IEXQ42GB-12--A| Item: 6482275

High dynamic binding capacity—ideal for capturing large proteins and viruses

$226.90CAD / Pkg of 4

$226.90CAD / Pkg of 4

Buy It Together

Current Item

Sartorius Sartobind® Lab Q Anion Exchange Membrane Adsorber, 0.41 mL; 4/PK
Sartorius Sartobind® Lab Q Anion Exchange Membrane Adsorber, 0.41 mL; 4/PK
6482275

$226.90CAD / Pkg of 4
+
Cole-Parmer Disposable Syringes, Luer-Lok®, 10 mL; 100/Pk
1291502

$120.67CAD / Pkg of 100
=
Total Price 347.57 ADD 2 ITEMS TO CART
Product Description

Includes

One male Luer and one female Luer PEEK adapters to UNF 10-32.

Key Features

  • Ready-to-use reusable adsorbers optimize cost efficiency
  • Connect in series for increased binding capacity
  • Lab IEX 15 membrane adsorber features a 15 cm² membrane area with 3 layers and 25 mm membrane diameter

More About this Item

Sartorius Sartobind® Lab membrane adsorbers for ion exchange chromatography are ready-to-use units in a syringe filter format. They are based on the patented macroporous Sartobind membranes, with anion or cation exchange ligands covalently bound to the entire internal surface of the pores. This results in a chromatography matrix with superior flow rates and high binding capacity. Covalent linkage of ion exchange ligands to the membrane is very stable, allowing for many cycles of re-use without loss of binding capacity. Equipped with standard Luer lock connectors, Sartobind Lab units enable simple, equipment-free handling by syringe, or can be easily connected to FPLC systems for automated purifications. The units are compatible with most common chromatography buffers and can be re-used > 1,000 times, ensuring high-cost efficiency.

Sartobind Lab units for ion exchange are available with strong (Q) or weak (D) anion and strong cation (S) exchangers, and in three bed volumes, 0.41, 2.1, and 2.8 mL, corresponding to dynamic binding capacities between 10.5 and 80 mg protein per unit.

Specifications
  • Membrane MaterialStabilized reinforced cellulose
  • Pore Size (micron)3
  • Filter Size25 mm Diameter Membrane
  • Housing MaterialPolypropylene
  • ConnectionsFemale Luer Inlet x Male Luer Outlet
  • Capacity (mL)0.41
  • Effective Filtration Area (cm²)15
  • Typical Water Flow Rate2 to 12 mL/min
  • Max Pressure (PSI)87
  • DescriptionSartobind® Lab Q Anion Exchange Membrane Adsorber, 0.41 mL; 4/PK
Reviews
Q&A