ResinTech VP-Series Water Purification Cartridges
Reduce operating costs while maintaining peak performance
  • High-capacity cartridges require fewer changes
  • Spin-welded construction with high-purity plastics ensures long service life
  • Available in standard connection and hose nipple configurations
  • Made in the USA!
Use these filtration cartridges in your ResinTech, UNIX, Barnstead® PCS® and Vaponics® water purification systems or design a system using wall-mounting brackets and extension tubing (order separately). Permanent joint welds create a hermetic seal and improve longevity in any installation. Nonpigmented ABS plastic housing is durable and produces high-purity laboratory-grade water. Choose between the standard ARIES connection type and the versatile 3/8" hose nipple, both of which are available in a variety of filter media.
$227.20 - $350.08CAD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Compatible with Media
Availability Pricing
ResinTech VP-17-4030 VP Series High Purity Color-Changing DI Filter Cartridge (Std.) 9928473 Mfr # VP-17-4030
COMPARE
1360 grains ARIES, Unix, Barnstead, Vaponics High Purity, Mixed Bed, Color Changing
$300.50CAD / Each
ResinTech VP-17-4040 VP Series High Capacity Color-Changing DI Filter Cartridge (Std.) 9928474 Mfr # VP-17-4040
COMPARE
1590 grains ARIES, Unix, Barnstead, Vaponics High Capacity, Mixed Bed, Color Changing
$300.50CAD / Each
ResinTech VP-17-4010 VP Series High Purity DI Filter Cartridge (Std.) 9928476 Mfr # VP-17-4010
COMPARE
1360 grains ARIES, Unix, Barnstead (D0809, D5027, D8816), Vaponics (MR-1, MR-2) High Purity, Mixed-Bed
$277.96CAD / Each
ResinTech VP-17-4011 VP Series Low TOC DI Filter Cartridge (Std.) 9928477 Mfr # VP-17-4011
COMPARE
1360 grains ARIES, Unix, Barnstead (D0820, D5021), Vaponics (MR-1-LTOC, MR-2-LTOC) Low TOC, Mixed-Bed
$336.74CAD / Each
ResinTech VP-17-1050 VP Series Activated Carbon Filter Cartridge (Std.) 9928479 Mfr # VP-17-1050
COMPARE
2000 gal. ARIES, Unix, Barnstead (D0813), Vaponics (AC-1, AC-2) Activated Carbon
$242.00CAD / Each
ResinTech VP-17-3680 VP Series Oxygen Reduction Filter Cartridge (Std.) 9928480 Mfr # VP-17-3680
COMPARE
1980 ppm ARIES, Unix, Barnstead (D0811, D8817), Vaponics (OX-1, OX-2) Oxygen Removal
$350.08CAD / Each
ResinTech VP-17-4210 VP Series Organics Removal Dual Bed 9928481 Mfr # VP-17-4210
COMPARE
1008 grains ARIES, Unix, Barnstead (D0835), Vaponics Dual Bed, Activated Carbon, Macropore Anion
$296.67CAD / Each
ResinTech VP-17-4205 VP Series High Purity Organics Filter Cartridge (Std.) 9928482 Mfr # VP-17-4205
COMPARE
490 grains / 2000 gal. ARIES, Unix, Barnstead (D0832), Vaponics (MRO-1, MRO-2) Blended, High Purity, Activated Carbon
$251.21CAD / Each
ResinTech VP-17-4200 VP Series High Purity Oxygen Reduction Filter Cartridge (Std.) 9928483 Mfr # VP-17-4200
COMPARE
400 grains / 660 ppm ARIES, Unix, Barnstead (D8809), Vaponics (MRX-1, MRX-2) Blended, High Purity, Oxygen Removal
$331.44CAD / Each
ResinTech VP-17-4301 VP Series High Purity/Oxygen/Organics Filter Cartridge (Std.) 9928484 Mfr # VP-17-4301
COMPARE
300 grains / 525 ppm ARIES, Unix, Barnstead (D8811, D8818), Vaponics (MROX-1, MROX-2) Triple Layer, High Purity, Activated Carbon, Oxygen Removal
$318.03CAD / Each
ResinTech VP-17-4203 VP Series High Capacity Filter Cartridge (Std.) 9928485 Mfr # VP-17-4203
COMPARE
1590 grains ARIES, Unix, Barnstead (D0803), Vaponics Dual Layer, High Capacity, Anion, Cation
$275.23CAD / Each
ResinTech VP-17-4010-HN VP Series High Purity DI Filter Cartridge, Hose Nipple 9928486 Mfr # VP-17-4010-HN
COMPARE
1360 grains ARIES, Unix, Barnstead (D8902, D8911), Vaponics (MRN-1) High Purity, Mixed-Bed
$277.96CAD / Each
ResinTech VP-17-4030-HN VP Series Color Changing High Purity Mixed Bed, Hose Nipple 9928487 Mfr # VP-17-4030-HN
COMPARE
1360 grains ARIES, Unix, Barnstead, Vaponics High Purity, Mixed Bed, Color Changing
$300.50CAD / Each
ResinTech VP-17-1050-HN VP Series Organics Removal, Hose Nipple 9928489 Mfr # VP-17-1050-HN
COMPARE
2000 gal. ARIES, Unix, Barnstead, Vaponics Activated Carbon
$227.20CAD / Each
ResinTech VP-17-0001 VP Series Sanitization Cartridge 9928530 Mfr # VP-17-0001
COMPARE
$242.37CAD / Each
Showing 1- 15 of 15