QLA Hanson-Compatible Sampling Probes and Cannulas
Achieve consistent sampling with QLA Hanson-compatible probes and cannulas
  • Matching cannula to stopper ensures greater sampling position consistency
For dissolution tests, taking reliable samples is critical. QLA probes, cannula, and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
$94.74 - $383.45CAD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (") Material Description
Availability Pricing
QLA Bent PEEK Cannula for Hanson; Custom 450 mm Length 4130225 Mfr # CAN450MM-PH
COMPARE
17 45/64 PEEK Bent PEEK Cannula for Hanson; Custom 450 mm Length
$275.52CAD / Each
QLA Bent 316 SS Cannula for Hanson; Custom 450 mm Length 4130226 Mfr # CAN450MM-SH
COMPARE
17 45/64 316 stainless steel Bent 316 SS Cannula for Hanson; Custom 450 mm Length
$291.36CAD / Each
QLA Bent 316 SS Cannula for Hanson, 4.75"L, 1/16" OD; PVDF Luer Lock 4130229 Mfr # CAN475-062L
COMPARE
4 3/4 316 SS cannula/PVDF luer lock Bent 316 SS Cannula for Hanson, 4.75"L, 1/16" OD; PVDF Luer Lock
$94.74CAD / Each
QLA Bent 316 SS Cannula for Hanson, 4.75"L; SS Luer Lock and Perm SS Tip 4130230 Mfr # CAN475-HR
COMPARE
4 3/4 316 stainless steel Bent 316 SS Cannula for Hanson, 4.75"L; SS Luer Lock and Perm SS Tip
$250.11CAD / Each
QLA Bent 316 SS Cannula for Hanson, 4.75", 1/8" OD; PVDF Luer Lock 4130234 Mfr # CAN475-SHL
COMPARE
4 49/64 316 SS cannula/PVDF luer lock Bent 316 SS Cannula for Hanson, 4.75", 1/8" OD; PVDF Luer Lock
$120.23CAD / Each
QLA Dual DV Series Sampling Probe for Hanson SR8-Plus; for 500 mL Vessel 4130238 Mfr # CAN500-DHV
COMPARE
Dual DV Series Sampling Probe for Hanson SR8-Plus; Use with 500 mL vessel
$351.77CAD / Each
QLA Bent 316 SS Cannula for Hanson, 7.75"L, 1/16" OD; PVDF Luer Lock 4130247 Mfr # CAN775-062L
COMPARE
7 3/4 316 SS cannula/PVDF luer lock Bent 316 SS Cannula for Hanson, 7.75"L, 1/16" OD; PVDF Luer Lock
$113.01CAD / Each
QLA Bent 316 SS Cannula for Hanson, 7.75"L; SS Luer Lock and Perm SS Tip 4130248 Mfr # CAN775-HR
COMPARE
7 3/4 316 stainless steel Bent 316 SS Cannula for Hanson, 7.75"L; SS Luer Lock and Perm SS Tip
$383.45CAD / Each
QLA Bent 316 SS Cannula for Hanson, 7.75"L, 1/8" OD; PVDF Luer Lock 4130251 Mfr # CAN775-SHL
COMPARE
7 3/4 316 SS cannula/PVDF luer lock Bent 316 SS Cannula for Hanson, 7.75"L, 1/8" OD; PVDF Luer Lock
$154.34CAD / Each
QLA Standard Sampling Probe for Hanson/"HR" filters; Use with 900 mL vessel 4130255 Mfr # CAN900-DH
COMPARE
Standard Sampling Probe for Hanson/"HR" filters; Use with 900 mL vessel
$254.90CAD / Each
QLA Dual DV Series Sampling Probe for Hanson SR8-Plus; for 900 mL Vessel 4130256 Mfr # CAN900-DHV
COMPARE
Dual DV Series Sampling Probe for Hanson SR8-Plus; Use with 900 mL vessel
$351.77CAD / Each
QLA Straight PEEK Cannula for Hanson, 15", 1/8" OD; PEEK Luer Lock 4130265 Mfr # CANS15-PH
COMPARE
15 PEEK Straight PEEK Cannula for Hanson, 15", 1/8" OD; PEEK Luer Lock
$180.49CAD / Each
QLA Straight 316 SS Cannula for Hanson, 15", 1/8" OD; SS Luer Lock 4130266 Mfr # CANS15-SH
COMPARE
15 316 stainless steel Straight 316 SS Cannula for Hanson, 15", 1/8" OD; SS Luer Lock
$160.23CAD / Each
Showing 1- 13 of 13