QLA Standard Sampling Probe for Hanson/"HR" filters; Use with 900 mL vessel

Representative image only.

QLA Standard Sampling Probe for Hanson/"HR" filters; Use with 900 mL vessel

| Mfr: CAN900-DH| Item: 4130255

Achieve consistent sampling with QLA Hanson-compatible probes and cannulas

$254.90CAD / Each

$254.90CAD / Each

Buy It Together

Current Item

QLA Standard Sampling Probe for Hanson/"HR" filters; Use with 900 mL vessel
QLA Standard Sampling Probe for Hanson/"HR" filters; Use with 900 mL vessel
4130255

$254.90CAD / Each
+ =
Total Price 368.65 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Matching cannula to stopper ensures greater sampling position consistency
  • Standard sampling probe
  • Use with 900 ml vessel

More About this Item

For dissolution tests, taking reliable samples is critical. QLA probes, cannula, and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.

.

Specifications
  • DescriptionStandard Sampling Probe for Hanson/"HR" filters; Use with 900 mL vessel
Reviews
Q&A