PolyScience Refrigerated Circulating Baths

Advanced PID algorithms increase heat/cool responsiveness to ensure constant temperature
  • Waterproof control panel swivels 180°
  • Extra-large backlit digital display can be easily seen from across a room
The unique WhisperCool™ environmental control system features part-load sound levels below that of conventional speech at a distance of two meters. This innovative environmental technology also optimizes compressor and evaporator performance, reducing overall energy consumption and prolonging compressor life. The front-mounted drain port and washable air filter simplify cleaning and maintenance. The chemical-resistant DuraTop™ top plate remains cool at high temperatures and is easily cleaned. Rear inlet/outlet ports also allow use in external process applications.
$10,792.58 - $13,363.65CAD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
PolyScience PD15R-40-A11B Performance Digital Recirculating Bath, 15L, -40 to 200 °C, 120V/60Hz 1221026 Mfr # AP15R-40-C11B
COMPARE
200 -40 Performance Digital 15L Recirculating Bath, -40 to 200 °C, 120V/60Hz
$13,010.03CAD / Each
PolyScience SD28R-30-A11B Standard Digital Recirculating Bath, 28L, -30 to 170 °C, 120V/60Hz 1221036 Mfr # SD28R-30-A11B
COMPARE
170 -30 Standard Digital 28L Refrigerated Bath Circulator, -30 to 170 °C, 120V/60Hz
$10,792.58CAD / Each
PolyScience PD28R-30-A11B Performance Digital Recirculating Bath, 28L, -30 to 200 °C, 120V/60Hz 1221038 Mfr # AP28R-30-A11B
COMPARE
200 -30 Performance Digital 28L Recirculating Bath, -30 to 200 °C, 120V/60Hz
$13,363.65CAD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat