Polyscience Air-Cooled Recirculator
Ideal for applications where cooling fluid temperature is higher than ambient temperature
This air-cooled recirculator is an economical alternative to chillers with mechanical refrigeration. A positive displacement pump circulates fluid to your equipment, into a reservoir, and then through a fan-cooled radiator that removes the heat. Built-in low liquid level indicator ensures the safety of the reservoir and pumping system. Compact design saves space in your laboratory or factory.
$3,867.65 - $4,287.56CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (Hz) Power (VAC)
Availability Pricing
PolyScience Air-Cooled Recirculator; 120V; 60Hz; 5.5A 1293000 Mfr # 3370P9C11B
COMPARE
60 115
$4,287.56CAD / Each
PolyScience Air-Cooled Recirculator; 240V; 50Hz; 3A 1293005 Mfr # 3370P9A12E
COMPARE
50 230
$3,867.65CAD / Each