Plate Mill
Heavy-duty plate mills provide clog and heat-resistant operation
  • Low output speed makes mill ideal for grinding hard and dry materials
  • Grinding unit disassembles without tools for easy cleaning
Use this heavy-duty plate mill for a variety of grinding applications: food, pharmaceutical, chemical, soil testing, and ore and coal sampling. Grinding unit is constructed of cast iron and coated with lead-free tin for food handling applications. Adjust the thumbscrew on the grinding unit to change the plate gap from 1/4" open to nearly closed. Includes grinding plates for fine grinding; for coarse grinding, order optional coarse grinding plate set 04146-53 separately. Grinding plates and feeds are made of durable iron alloy. Use the tooth feed (included) for dry materials and the worm feed (optional) for wet or oily materials.
$5,289.47 - $6,018.80CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Plate mill, with standard motor, 115 VAC, 60 Hz 0418600 Mfr # S-4E-1-6TB
COMPARE
115
$5,289.47CAD / Each
Plate mill, with standard motor, 220 VAC, 50 Hz 0418605 Mfr # S-4E-2-5/6TB
COMPARE
230
$6,018.80CAD / Each