Pens for Barton, American Meter, and Mercury Recorders
Get exacting performance that meets American Gas Association (AGA) standards
  • Static pen's intense black ink reduces time spent retracing
  • Bias-cut pen nib covering provides enhanced visibility while arcing and setting time lags
  • All-temperature ink ensures a solid trace in all climates
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Scannable pens produce broader lines for reliable scanner processing
  • Fine-point pens produce thinner lines allowing integrator operators to trace gas flows more accurately
$172.39CAD / Pkg of 6
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Pen Color Tip Style Description
Availability Pricing
Graphic Controls 82-79-0312-06 Scannable Pen, Barton, red differential RK-49000-26 Mfr # 82-79-0312-06
COMPARE
Red Broad-point Scannable Pen, Barton, red differential
$172.39CAD / Pkg of 6
Graphic Controls 82-72-0213-06 Scannable Pen, American Meter/Mercury, blue static RK-49000-31 Mfr # 82-72-0213-06
COMPARE
Blue Broad-point Scannable Pen, American Meter/Mercury, blue static
$172.39CAD / Pkg of 6
Graphic Controls 82-72-0114-06 Scannable Pen, American Meter/Mercury, green temperature RK-49000-32 Mfr # 82-72-0114-06
COMPARE
Green Broad-point Scannable Pen, American Meter/Mercury, green temperature
$172.39CAD / Pkg of 6
Graphic Controls 82-79-3312-06 Fine Line Pen, Barton, red differential RK-49000-34 Mfr # 82-79-3312-06
COMPARE
Red Fine-point Fine Line Pen, Barton, red differential
$172.39CAD / Pkg of 6
Graphic Controls 82-72-3113-06 Fine Line Pen, Barton, blue static RK-49000-36 Mfr # 82-72-3113-06
COMPARE
Blue Fine-point Fine Line Pen, Barton, blue static
$172.39CAD / Pkg of 6
Graphic Controls 82-79-3412-06 Fine Line Pen, American Meter/Mercury, red differential RK-49000-37 Mfr # 82-79-3412-06
COMPARE
Red Fine-point Fine Line Pen, American Meter/Mercury, red differential
$172.39CAD / Pkg of 6
Graphic Controls 82-72-3114-06 Fine Line Pen, American Meter/Mercury, green temp RK-49000-40 Mfr # 82-72-3114-06
COMPARE
Green Fine-point Fine Line Pen, American Meter/Mercury, green temp
$172.39CAD / Pkg of 6
Showing 1- 7 of 7