Graphic Controls 82-72-0114-06 Scannable Pen, American Meter/Mercury, green temperature
Representative image only.
Graphic Controls 82-72-0114-06 Scannable Pen, American Meter/Mercury, green temperature
Graphic Controls | Mfr: 82-72-0114-06| Item: 4900032
Get exacting performance that meets American Gas Association (AGA) standards
$172.39CAD / Pkg of 6
$172.39CAD / Pkg of 6
Product Description
Key Features
  • Static pen's intense black ink reduces time spent retracing
  • Bias-cut pen nib covering provides enhanced visibility while arcing and setting time lags
  • All-temperature ink ensures a solid trace in all climates
  • Calibrate pen arrays to a 15-minute time lag between static and differential on the Zero Line of a 24 hour chart
  • Scannable pens produce broader lines for reliable scanner processing
  • Fine-point pens produce thinner lines allowing integrator operators to trace gas flows more accurately
  • 1 year warranty
1 Year Warranty
Specifications
  • Tip StyleBroad-point
  • Pen ColorGreen
  • DescriptionScannable Pen, American Meter/Mercury, green temperature