Ovation M Mechanical Adjustable-Volume-Set Pipettors
Ergonomics and performance with innovation you can feel
  • Easily contours to your hand for ultimate comfort
  • Only minimal force is required in the proven ergonomic design
  • Built-in cap opener at base opens PCR and microtubes with ease
With value in mind, these mechanical adjustable-volume pipettors use the same proven ergonomic Ovation design to minimize contact pressure, tension, and exertion.
The low force plunger allows you to manually control speed with a light touch of the thumb. Releasing pipette tips is a breeze with just a quick press of the tip eject button. The one-way nozzle protects your pipettor and samples as it blocks access of liquid into the body of the pipettor.
For maximum efficiency, use the built-in cap opener at the base of the pipettor to open tops of PCR and microtubes. No pipette stands are necessary, the Ovation pipettor stands by itself on the benchtop. Order the optional stand to hold up to four pipettors at a time, saving you valuable bench space in the lab.
$609.98CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Description
Availability Pricing
VistaLab Ovation M M Mechanical Volume Set Pipette, Orange, 1 to 10 uL 0619904 Mfr # 1070-0010
COMPARE
10 1 M Mechanical Volume Set Pipette, Orange, 1 to 10 uL
$609.98CAD / Each
VistaLab Ovation M M Mechanical Volume Set Pipette, Green, 10 to 100 uL 0619908 Mfr # 1070-0100
COMPARE
100 10 M Mechanical Volume Set Pipette, Green, 10 to 100 uL
$609.98CAD / Each
VistaLab Ovation M M Mechanical Volume Set Pipette, Blue, 20 to 200 uL 0619910 Mfr # 1070-0200
COMPARE
200 20 M Mechanical Volume Set Pipette, Blue, 20 to 200 uL
$609.98CAD / Each
VistaLab Ovation M M Mechanical Volume Set Pipette, Purple, 100 to 1000 uL 0619912 Mfr # 1070-1000
COMPARE
1000 100 M Mechanical Volume Set Pipette, Purple, 100 to 1000 uL
$609.98CAD / Each