VistaLab Ovation M M Mechanical Volume Set Pipette, Orange, 1 to 10 uL
Representative image only.
VistaLab Ovation M M Mechanical Volume Set Pipette, Orange, 1 to 10 uL
VistaLab | Mfr: 1070-0010| Item: 0619904
Ergonomics and performance with innovation you can feel
$609.98CAD / Each
$609.98CAD / Each
Buy It Together
Current Item
VistaLab Ovation M M Mechanical Volume Set Pipette, Orange, 1 to 10 uL
VistaLab Ovation M M Mechanical Volume Set Pipette, Orange, 1 to 10 uL
0619904
$609.98CAD / Each
+ =
Total Price 744.80 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Easily contours to your hand for ultimate comfort
  • Only minimal force is required in the proven ergonomic design
  • Built-in cap opener at base opens PCR and microtubes with ease
  • 1 year warranty
More About this Item
With value in mind, these mechanical adjustable-volume pipettors use the same proven ergonomic Ovation design to minimize contact pressure, tension, and exertion.
The low force plunger allows you to manually control speed with a light touch of the thumb. Releasing pipette tips is a breeze with just a quick press of the tip eject button. The one-way nozzle protects your pipettor and samples as it blocks access of liquid into the body of the pipettor.
For maximum efficiency, use the built-in cap opener at the base of the pipettor to open tops of PCR and microtubes. No pipette stands are necessary, the Ovation pipettor stands by itself on the benchtop. Order the optional stand to hold up to four pipettors at a time, saving you valuable bench space in the lab.
1 Year WarrantyCanadian Standards AssociationETL ListedISO9001 Registered SupplierUL Listed.
Specifications
  • SterileNo
  • Max Volume (µL)10
  • Min Volume (µL)1
  • DescriptionM Mechanical Volume Set Pipette, Orange, 1 to 10 uL