Orochem Reliasil™ C4 HPLC Columns

Rugged design with enhanced chemical stability and efficiency
  • Excellent mechanical and chemical stability
  • Ideal for reverse phase separations of peptides, pharmaceuticals, and derivatized amino acids
  • Suitable for C4 phase
The Orochem Reliasil HPLC columns are designed for high efficiency (high plate count) and general utility. The proprietary silica gel manufacturing process yields a Gaussian particle size distribution, narrow pore-size distribution, and strong resistance to high pH. All columns feature a 190 m²/g surface area and 0.55 pore volume. Choose from standard columns with a pore size of 90 Å or columns with a 300 Å pore size.
$1,012.22 - $1,390.88CAD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Description
Availability Pricing
Orochem Reliasil™ HPLC Column, C4, 5 µm, 100 mm L x 3.0 mm ID, 90Å 0648654 Mfr # R5DG-123
COMPARE
3 100 Reliasil HPLC Column, C4, 100 x 3 mm, 5 µm
$1,012.22CAD / Each
Orochem Reliasil™ HPLC Column, C4, 5 µm, 150 mm L x 4.6 mm ID, 300Å 0648655 Mfr # R5FI-128
COMPARE
4.6 150 Reliasil HPLC Column, C4, 150 x 4.6 mm, 5 µm, 300 Å
$1,237.65CAD / Each
Orochem Reliasil™ HPLC Column, C4, 5 µm, 150 mm L x 4.6 mm ID, 90Å 0648656 Mfr # R5FI-123
COMPARE
4.6 150 Reliasil HPLC Column, C4, 150 x 4.6 mm, 5 µm
$1,046.11CAD / Each
Orochem Reliasil™ HPLC Column, C4, 5 µm, 250 mm L x 4.6 mm ID, 300Å 0648657 Mfr # R5HI-128
COMPARE
4.6 250 Reliasil HPLC Column, C4, 250 x 4.6 mm, 5 µm, 300 Å
$1,390.88CAD / Each
Orochem Reliasil™ HPLC Column, C4, 5 µm, 250 mm L x 4.6 mm ID, 90Å 0648658 Mfr # R5HI-123
COMPARE
4.6 250 Reliasil HPLC Column, C4, 250 x 4.6 mm, 5 µm
$1,196.39CAD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat