Orochem Reliasil™ HPLC Column, C4, 5 µm, 250 mm L x 4.6 mm ID, 300Å

Representative image only.

Orochem Reliasil™ HPLC Column, C4, 5 µm, 250 mm L x 4.6 mm ID, 300Å

Orochem | Mfr: R5HI-128| Item: 0648657

Rugged design with enhanced chemical stability and efficiency

$1,390.88CAD / Each

$1,390.88CAD / Each
Product Description

Key Features

  • Excellent mechanical and chemical stability
  • Ideal for reverse phase separations of peptides, pharmaceuticals, and derivatized amino acids
  • Suitable for C4 phase
  • C4 column dimensions are 250 mm long by 5 mm diameter

More About this Item

The Orochem Reliasil HPLC columns are designed for high efficiency (high plate count) and general utility. The proprietary silica gel manufacturing process yields a Gaussian particle size distribution, narrow pore-size distribution, and strong resistance to high pH. All columns feature a 190 m²/g surface area and 0.55 pore volume. Choose from standard columns with a pore size of 90 Å or columns with a 300 Å pore size.

Specifications
  • PhaseC4
  • Column Length (mm)250
  • Column Inside Diameter (ID) (mm)4.6
  • Column Particle Size (µm)5
  • DescriptionReliasil HPLC Column, C4, 250 x 4.6 mm, 5 µm, 300 Å
Reviews
Q&A