Orochem Reliasil™ Amino HPLC Columns
Rugged design with enhanced chemical stability and efficiency
  • Excellent mechanical and chemical stability
  • Ideal for reverse phase separations of peptides, pharmaceuticals, and derivatized amino acids
  • Suitable for Amino phase
The Orochem Reliasil HPLC columns are designed for high efficiency (high plate count) and general utility. The proprietary silica gel manufacturing process yields a Gaussian particle size distribution, narrow pore-size distribution, and strong resistance to high pH. All columns feature a 190 m²/g surface area and 0.55 pore volume.Standard columns feature a pore size of 90 Å.
$1,012.22 - $9,577.04CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Orochem Reliasil™ HPLC Column, Amino, 3 µm, 100 mm L x 3.0 mm ID 0648634 Mfr # R3DG-103
COMPARE
3 100 3
$1,091.78CAD / Each
Orochem Reliasil™ HPLC Column, Amino, 5 µm, 100 mm L x 3.0 mm ID 0648635 Mfr # R5DG-132
COMPARE
3 100 5
$1,012.22CAD / Each
Orochem Reliasil™ HPLC Column, Amino, 3 µm, 150 mm L x 4.6 mm ID 0648636 Mfr # R3FI-103
COMPARE
4.6 150 3
$1,046.11CAD / Each
Orochem Reliasil™ HPLC Column, Amino, 3 µm, 150 mm L x 4.6 mm ID 0648637 Mfr # R5FI-132
COMPARE
4.6 150 5
$1,046.11CAD / Each
Orochem Reliasil™ HPLC Column, Amino, 3 µm, 250 mm L x 4.6 mm ID 0648638 Mfr # R5HI-132
COMPARE
4.6 250 5
$1,196.39CAD / Each
Orochem Reliasil™ HPLC Column, Amino, 10 µm, 250 mm L x 10 mm ID 0648639 Mfr # R10HL-142
COMPARE
10 250 10
$5,274.74CAD / Each
Orochem Reliasil™ HPLC Column, Amino, 3 µm, 250 mm L x 10 mm ID 0648640 Mfr # R9HL-142
COMPARE
10 250 5
$2,276.39CAD / Each
Orochem Reliasil™ HPLC Column, Amino, 10 µm, 250 mm L x 21 mm ID 0648641 Mfr # R10HT-142
COMPARE
21 250 10
$9,577.04CAD / Each
Orochem Reliasil™ HPLC Column, Amino, 3 µm, 250 mm L x 21 mm ID 0648642 Mfr # R9HM-142
COMPARE
21 250 5
$5,635.72CAD / Each
Showing 1- 9 of 9