Orochem Reliasil™ HPLC Column, Amino, 10 µm, 250 mm L x 10 mm ID

Representative image only.

Orochem Reliasil™ HPLC Column, Amino, 10 µm, 250 mm L x 10 mm ID

Orochem | Mfr: R10HL-142| Item: 0648639

Rugged design with enhanced chemical stability and efficiency

$5,274.74CAD / Each

$5,274.74CAD / Each
Product Description

Key Features

  • Excellent mechanical and chemical stability
  • Ideal for reverse phase separations of peptides, pharmaceuticals, and derivatized amino acids
  • Suitable for Amino phase
  • Amino column dimensions are 250 mm long by 10 mm diameter

More About this Item

The Orochem Reliasil HPLC columns are designed for high efficiency (high plate count) and general utility. The proprietary silica gel manufacturing process yields a Gaussian particle size distribution, narrow pore-size distribution, and strong resistance to high pH. All columns feature a 190 m²/g surface area and 0.55 pore volume.Standard columns feature a pore size of 90 Å.

Specifications
  • PhaseAmino
  • Column Length (mm)250
  • Column Inside Diameter (ID) (mm)10
  • Column Particle Size (µm)10
  • DescriptionReliasil HPLC Column, Amino, 250 x 10 mm, 10 µm
Reviews
Q&A