Orochem Epitomize™ Chiral Phase Columns
Chiral stationary phase columns with proven results and reproducibility
  • Individually tested to guarantee your results
  • Chiral stationary phase columns ideal for UHPLC, HPLC, Preparative, and SMB applications
  • Coated polysaccharides onto modified silica gel
These Orochem Epitomize™ CSP-1 series chiral stationary phase columns are based on derivatized polysaccharides that are coated onto modified silica gel. Recommended mobile phases are blends of alcohols and hydrocarbons at all rations and acetonitrile. Each column is tested before packaging and a chromatogram with test conditions and column performance data is included.
$2,593.17 - $4,287.56CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (mm) Column Particle Size (µm)
Availability Pricing
Orochem Epitomize™ Chiral Stationary Phase Column, CSP-1A, 100 x 3 mm, 3 µm 0648600 Mfr # E3DG-2003
COMPARE
3 100 3
$2,593.17CAD / Each
Orochem Epitomize™ Chiral Stationary Phase Column, CSP-1A, 150 x 4.6 mm, 3 µm 0648601 Mfr # E3FI-2003
COMPARE
4.6 150 3
$3,278.29CAD / Each
Orochem Epitomize™ Chiral Stationary Phase Column, CSP-1A, 250 x 4.6 mm, 5 µm 0648602 Mfr # E5HI-2005
COMPARE
4.6 250 5
$4,287.56CAD / Each
Orochem Epitomize™ Chiral Stationary Phase Column, CSP-1C, 100 x 3 mm, 3 µm 0648603 Mfr # E3DG-1003
COMPARE
3 100 3
$2,593.17CAD / Each
Orochem Epitomize™ Chiral Stationary Phase Column, CSP-1C, 250 x 4.6 mm, 5 µm 0648607 Mfr # E5HI-1005
COMPARE
4.6 250 5
$4,287.56CAD / Each
Orochem Epitomize™ Chiral Stationary Phase Column, CSP-1Z, 100 x 3 mm, 3 µm 0648609 Mfr # E3DG-3003
COMPARE
3 100 3
$2,593.17CAD / Each
Orochem Epitomize™ Chiral Stationary Phase Column, CSP-1Z, 150 x 4.6 mm, 3 µm 0648610 Mfr # E3FI-3003
COMPARE
4.6 150 3
$3,278.29CAD / Each
Orochem Epitomize™ Chiral Stationary Phase Column, CSP-1Z, 250 x 4.6 mm, 5 µm 0648611 Mfr # E5HI-3005
COMPARE
4.6 250 5
$4,287.56CAD / Each
Orochem Epitomize™ Chiral Stationary Phase Column, CSP-1C, 150 x 4.6 mm, 3 µm 0684605 Mfr # E3FI-1003
COMPARE
4.6 150 3
$3,278.29CAD / Each
Showing 1- 9 of 9