Ohaus Ranger 3000 Compact Bench Scale

Increase productivity with rapid stabilization and large backlit display
  • Rapid stabilization produces accurate results within one second and eliminates your wait time
  • SmarText™ menu navigation eliminates the need to master complex commands
  • User-friendly keys dedicated to TARE, Yes, No, Back and Exit menu functions
  • Easy-to-use weigh-below hook offers the functionality to weigh items that cannot be easily lifted on to the weighing pan
  • RS-232 standard for connection to printer or PC
  • Integrated carrying handle and internal rechargeable battery for portability
  • Maximum display resolution of 1:30,000, and a certified resolution of 1:3,000
  • Weighing functions include percent, parts counting, check weighing, accumulation and dynamic weighing
The multipurpose Ranger 3000 has all of the capabilities to produce precise results rapidly for a variety of applications. With seven application modes, Ranger 3000 is the ideal scale to meet many industrial weighing needs. Portable and equipped with a rechargeable battery, this scale is beneficial in every corner of the factory.

Standard features include weigh-below hook, integral transportation handle, sealed front panel, menu lock switch, up front level indicator, adjustable leveling feet, selectable environmental and auto-print settings, stability indicator, overload and underload indicators, low battery indicator, auto shut-off and auto tare.
$1,511.70CAD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Ohaus R31P15 Ranger 3000 Compact Bench Scale 15kg x 0.5g 1161073 Mfr # R31P15
COMPARE
15 30 100 to 240
$1,511.70CAD / Each
Ohaus R31P1502 Ranger 3000 Compact Bench Scale 1.5kg x 0.05g 1161074 Mfr # R31P1502
COMPARE
1.5 3 100 to 240
$1,511.70CAD / Each
Ohaus R31P3 Ranger 3000 Compact Bench Scale 3kg x 0.1g 1161075 Mfr # R31P3
COMPARE
3 6 100 to 240
$1,511.70CAD / Each
Ohaus R31P30 Ranger 3000 Compact Bench Scale 30kg x 1g 1161076 Mfr # R31P30
COMPARE
30 66 100 to 240
$1,511.70CAD / Each
Ohaus R31P6 Ranger 3000 Compact Bench Scale 6kg x 0.2g 1161077 Mfr # R31P6
COMPARE
6 15 100 to 240
$1,511.70CAD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat