Ohaus® PS Handheld Balances
Unique and functional combination cover/pan slides back for weighing and slides forward for covering the keys and display
  • Compact size fits easily in your pocket
  • Quick and easy operation with two-button design
  • Built-in stability and over/under load indicators
  • Load-cell lock protects the weighing cell from damage during transport
  • Durable ABS front panel with carbon steel rear housing
$167.97 - $215.11CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Capacity (g) Readability (g)
Availability Pricing
Ohaus PS121 PS Handheld Balance, 120g x 0.1g (High Contrast Lcd) 1110089 Mfr # PS121
COMPARE
2 CR2032 Lithium (included) 120 0.1
$167.97CAD / Each
Ohaus PS251 PS Handheld Balance, 250g x 0.1g (High Contrast Lcd) 1111003 Mfr # PS251
COMPARE
2 CR2032 Lithium (included) 250 0.1
$215.11CAD / Each